siRNA Details
| Gene Symbol: CDT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030928.3 | NP_112190.2 | 10 | 1912 | ||
| GenBank mRNA | AB053172.1 | - | 10 | 1912 | ||
| AF070552.1 | - | 8 | 1451 | |||
| AF321125.1 | - | 10 | 1883 | |||
| BC000137.2 | - | 10 | 1874 | 3048353 | ||
| BC008676.1 | - | 10 | 1750 | |||
| BC008860.2 | - | 10 | 1805 | |||
| BC009410.1 | - | 10 | 1868 | 3613566 | ||
| BC014202.2 | - | 6 | 1173 | |||
| BC021126.1 | - | 8 | 1444 | |||
| BC049205.1 | - | 8 | 1516 | |||

CDT1
Genomic Map