siRNA Details
| Gene Symbol: CTC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025099.5 | NP_079375.3 | 23 | 4793 | ||
| NR_046431.1 | 22 | 4623 | ||||
| XM_006721577.3 | XP_006721640.1 | 22 | 4648 | |||
| XM_006721578.3 | XP_006721641.1 | 21 | 4546 | |||
| XM_011524010.2 | XP_011522312.1 | 23 | 4672 | |||
| XM_011524011.2 | XP_011522313.1 | 22 | 4542 | |||
| GenBank mRNA | AL831955.1 | - | 22 | 4570 | ||

CTC1
Genomic Map