siRNA Details
| Gene Symbol: BBS10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024685.3 | NP_078961.3 | 2 | 638 | ||
| GenBank mRNA | AK304178.1 | - | 2 | 611 | ||
| BC026355.1 | - | 2 | 623 | 4817227 | ||
| DR155669.1 | - | 2 | 582 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

BBS10| Gene Symbol: BBS10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024685.3 | NP_078961.3 | 2 | 638 | ||
| GenBank mRNA | AK304178.1 | - | 2 | 611 | ||
| BC026355.1 | - | 2 | 623 | 4817227 | ||
| DR155669.1 | - | 2 | 582 | |||