siRNA Details
| Gene Symbol: ROR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001318204.1 | NP_001305133.1 | 3 | 408 | ||
| NM_004560.3 | NP_004551.2 | 3 | 408 | |||
| XM_005252008.4 | XP_005252065.1 | 3 | 373 | |||
| XM_006717121.3 | XP_006717184.1 | 3 | 1030 | |||
| XM_017014762.1 | XP_016870251.1 | 3 | 2105 | |||
| XM_017014763.1 | XP_016870252.1 | 3 | 284 | |||
| GenBank mRNA | AB209154.1 | - | 5 | 677 | ||
| BC033697.1 | - | 3 | 395 | |||
| BC051273.1 | - | 3 | 220 | |||
| BC130522.1 | - | 3 | 296 | 40146553 | ||
| M97639.1 | - | 3 | 408 | |||

ROR2
Genomic Map