siRNA Details
| Gene Symbol: ROR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_004560.3 | NP_004551.2 | 9 | 2060 | ||
| XM_005252008.4 | XP_005252065.1 | 9 | 2025 | |||
| XM_005252009.3 | XP_005252066.1 | 3 | 766 | |||
| XM_006717121.3 | XP_006717184.1 | 9 | 2682 | |||
| XM_017014762.1 | XP_016870251.1 | 9 | 3757 | |||
| XM_017014763.1 | XP_016870252.1 | 9 | 1936 | |||
| GenBank mRNA | AB209154.1 | - | 11 | 2329 | ||
| BC033697.1 | - | 9 | 2047 | |||
| BC130522.1 | - | 9 | 1948 | 40146553 | ||
| M97639.1 | - | 9 | 2060 | |||

ROR2
Genomic Map