siRNA Details
| Gene Symbol: CASQ2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001232.3 | NP_001223.2 | 10 | 1232 | ||
| GenBank mRNA | AK129831.1 | - | 2 | 87 | ||
| AK129891.1 | - | 10 | 1207 | |||
| AK225959.1 | - | 10 | 1094 | |||
| AK293595.1 | - | 8 | 1103 | |||
| AK295502.1 | - | 8 | 928 | |||
| AK313041.1 | - | 10 | 1178 | |||
| BC022288.1 | - | 10 | 1141 | 4289141 | ||
| D55655.1 | - | 10 | 1077 | |||

CASQ2
Genomic Map