siRNA Details
| Gene Symbol: C11orf87 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207645.3 | NP_997528.2 | 2 | 705 | ||
| XM_011542817.2 | XP_011541119.1 | 2 | 809 | |||
| XM_011542818.2 | XP_011541120.1 | 2 | 687 | |||
| GenBank mRNA | AK303693.1 | - | 2 | 705 | ||
| BC035798.1 | - | 2 | 625 | 5735100 | ||
| BC068577.1 | - | 2 | 678 | 4818568 | ||

C11orf87
Genomic Map