siRNA Details
| Gene Symbol: TCTN3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001143973.1 | NP_001137445.1 | 5 | 875 | ||
| NM_015631.5 | NP_056446.4 | 7 | 1112 | |||
| XM_005269690.1 | XP_005269747.1 | 7 | 954 | |||
| XM_011539627.1 | XP_011537929.1 | 7 | 954 | |||
| XM_011539628.1 | XP_011537930.1 | 7 | 954 | |||
| GenBank mRNA | AK075357.1 | - | 7 | 898 | ||
| AK098295.1 | - | 6 | 1090 | |||
| AK128834.1 | - | 6 | 1812 | |||
| AK299141.1 | - | 5 | 659 | |||
| AL050022.1 | - | 7 | 896 | |||
| AY359022.1 | - | 7 | 876 | |||
| BC009494.2 | - | 4 | 272 | |||
| BC061582.1 | - | 7 | 783 | |||
| BC068449.1 | - | 7 | 1112 | 5298627 | ||
| DQ278872.1 | - | 7 | 1112 | |||

TCTN3
Genomic Map