siRNA Details
| Gene Symbol: FGD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_004463.2 | NP_004454.2 | 15 | 2968 | ||
| GenBank mRNA | AK226117.1 | - | 15 | 2758 | ||
| AK299635.1 | - | 13 | 1677 | |||
| BC034530.1 | - | 15 | 2480 | 4938457 | ||
| U11690.1 | - | 15 | 2964 | |||

FGD1
Genomic Map