siRNA Details
Gene Symbol: SRCAP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006662.2 | NP_006653.2 | 30, 31 | 6986 | ||
GenBank mRNA | AF143946.1 | - | 25, 26 | 6034 | ||
AK128030.1 | - | 30, 31 | 6986 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: SRCAP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006662.2 | NP_006653.2 | 30, 31 | 6986 | ||
GenBank mRNA | AF143946.1 | - | 25, 26 | 6034 | ||
AK128030.1 | - | 30, 31 | 6986 |