siRNA Details
| Gene Symbol: SEPN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020451.2 | NP_065184.2 | 6, 7 | 920 | ||
| NM_206926.1 | NP_996809.1 | 5, 6 | 818 | |||
| GenBank mRNA | AF166125.1 | - | 4, 5 | 509 | ||
| AJ306399.1 | - | 6, 7 | 920 | |||
| AK308457.1 | - | 4, 5 | 538 | |||
| BC021028.2 | - | 1, 2 | 122 | |||
| BQ217758.1 | - | 3, 4 | 375 | |||

SEPN1
Genomic Map