siRNA Details
| Gene Symbol: OPN1LW | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020061.5 | NP_064445.2 | 5 | 986 | ||
| GenBank mRNA | AW950066.1 | - | 2 | 350 | ||
| BM688032.1 | - | 2 | 214 | |||
| CR749814.1 | - | 5 | 944 | |||
| EL949562.1 | - | 3 | 495 | |||
More Information
Set Membership: |
|

OPN1LW
Genomic Map