siRNA Details
| Gene Symbol: EPB42 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000119.2 | NP_000110.2 | 1 | 378 | ||
| XM_011521349.2 | XP_011519651.1 | 2 | 1106 | |||
| XM_011521350.2 | XP_011519652.1 | 3 | 838 | |||
| XM_011521351.2 | XP_011519653.1 | 3 | 700 | |||
| XM_011521352.2 | XP_011519654.1 | 1 | 317 | |||
| XM_011521353.2 | XP_011519655.1 | 1 | 316 | |||
| GenBank mRNA | M30647.1 | - | 1 | 305 | ||
| M60298.1 | - | 1 | 537 | |||

EPB42
Genomic Map