siRNA Details
| Gene Symbol: SCARF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153334.6 | NP_699165.3 | 6 | 1190 | ||
| NM_182895.4 | NP_878315.2 | 6 | 1190 | |||
| XM_017029065.1 | XP_016884554.1 | 6 | 1153 | |||
| GenBank mRNA | AB024433.1 | - | 6 | 1156 | ||
| AB052951.1 | - | 6 | 1096 | |||
| AF522196.1 | - | 6 | 1190 | |||
| BC009326.2 | - | 3 | 273 | |||

SCARF2
Genomic Map