siRNA Details
| Gene Symbol: DNAH8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001206927.1 | NP_001193856.1 | 25 | 3385 | ||
| XM_011514318.2 | XP_011512620.1 | 25 | 3396 | |||
| XM_011514319.2 | XP_011512621.1 | 25 | 3396 | |||
| XM_011514320.2 | XP_011512622.1 | 23 | 3159 | |||
| XM_011514321.1 | XP_011512623.1 | 22 | 2743 | |||
| XM_017010325.1 | XP_016865814.1 | 25 | 3396 | |||
| XM_017010326.1 | XP_016865815.1 | 25 | 3396 | |||
| XM_017010327.1 | XP_016865816.1 | 25 | 3397 | |||
| GenBank mRNA | AF356519.1 | - | 23 | 2879 | ||

DNAH8
Genomic Map