siRNA Details
| Gene Symbol: NBEAL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015175.2 | NP_055990.1 | 38 | 6456 | ||
| XM_006713072.3 | XP_006713135.1 | 37 | 6372 | |||
| XM_011533533.2 | XP_011531835.1 | 38 | 6453 | |||
| XM_017006010.1 | XP_016861499.1 | 38 | 6453 | |||
| XM_017006011.1 | XP_016861500.1 | 38 | 6309 | |||
| XM_017006012.1 | XP_016861501.1 | 37 | 6372 | |||
| XM_017006013.1 | XP_016861502.1 | 38 | 6453 | |||
| XM_017006014.1 | XP_016861503.1 | 37 | 6228 | |||
| XM_017006015.1 | XP_016861504.1 | 36 | 6084 | |||
| XM_017006016.1 | XP_016861505.1 | 33 | 7030 | |||
| XM_017006017.1 | XP_016861506.1 | 15 | 2872 | |||
| GenBank mRNA | AB011112.3 | - | 24 | 4140 | ||
| AK131104.1 | - | 38 | 6296 | |||
| AK301269.1 | - | 3 | 519 | |||
| AY358455.1 | - | 8 | 1406 | |||
| BC060874.1 | - | 6 | 934 | |||

NBEAL2
Genomic Map