siRNA Details
| Gene Symbol: KCNJ18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_005276919.3 | XP_005276976.1 | 2 | 2725 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

KCNJ18| Gene Symbol: KCNJ18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_005276919.3 | XP_005276976.1 | 2 | 2725 | ||