siRNA Details
| Gene Symbol: SDHB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003000.2 | NP_002991.2 | 4 | 500 | ||
| GenBank mRNA | AK312056.1 | - | 4 | 362 | ||
| BC007840.2 | - | 4 | 500 | 4124737 | ||
| BF796275.1 | - | 4 | 348 | |||
| BU554163.1 | - | 2 | 90 | |||
| D10245.1 | - | 3 | 295 | |||
| DQ403007.1 | - | 4 | 322 | |||
| M32246.1 | - | 3 | 295 | |||
| U17248.1 | - | 4 | 483 | |||

SDHB
Genomic Map