siRNA Details
| Gene Symbol: GYS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001161587.1 | NP_001155059.1 | 9 | 1307 | ||
| NM_002103.4 | NP_002094.2 | 10 | 1499 | |||
| NR_027763.1 | 9 | 1317 | ||||
| GenBank mRNA | AK223579.1 | - | 10 | 1455 | ||
| AK291672.1 | - | 10 | 1455 | |||
| AK294349.1 | - | 5 | 415 | |||
| AK300369.1 | - | 10 | 1287 | |||
| AK302738.1 | - | 9 | 1273 | |||
| AK303712.1 | - | 8 | 1155 | |||
| BC002617.2 | - | 10 | 1438 | 3143019 | ||
| BC003182.1 | - | 9 | 1264 | 2958297 | ||
| BC007688.2 | - | 10 | 1425 | 3953692 | ||
| J04501.1 | - | 10 | 1418 | |||
| U32573.1 | - | 10 | 1400 | |||

GYS1
Genomic Map