siRNA Details
| Gene Symbol: C12orf57 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001301834.1 | NP_001288763.1 | 4 | 627 | ||
| NM_001301836.1 | NP_001288765.1 | 3 | 624 | |||
| NM_001301837.1 | NP_001288766.1 | 3 | 669 | |||
| NM_001301838.1 | NP_001288767.1 | 3 | 852 | |||
| NM_138425.3 | NP_612434.1 | 3 | 756 | |||
| NR_126035.1 | 3 | 765 | ||||
| GenBank mRNA | AV751928.1 | - | 507 | |||
| BC009925.2 | - | 3 | 533 | 3537206 | ||
| BP318750.2 | - | 360 | ||||

C12orf57
Genomic Map