siRNA Details
| Gene Symbol: CLCNKA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001042704.1 | NP_001036169.1 | 18 | 1980 | ||
| NM_001257139.1 | NP_001244068.1 | 17 | 1854 | |||
| NM_004070.3 | NP_004061.3 | 18 | 1983 | |||
| GenBank mRNA | AK225550.1 | - | 18 | 1935 | ||
| AK290814.1 | - | 18 | 1938 | |||
| AK298285.1 | - | 17 | 1833 | |||
| BC035373.1 | - | 22 | 2507 | |||
| BC048282.1 | - | 17 | 1908 | 5184165 | ||
| BC053869.1 | - | 19 | 2028 | 5187202 | ||
| Z30643.1 | - | 17 | 1925 | |||

CLCNKA
Genomic Map