siRNA Details
| Gene Symbol: IL17RA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001289905.1 | NP_001276834.1 | 7 | 817 | ||
| NM_014339.6 | NP_055154.3 | 7 | 817 | |||
| GenBank mRNA | BC011624.2 | - | 7 | 761 | 4053746 | |
| BX334547.2 | - | 7 | 820 | |||
| BX410574.2 | - | 7 | 733 | |||
| CT841520.1 | - | 7 | 740 | |||
| JF305973.1 | - | 7 | 684 | |||
| U58917.1 | - | 7 | 716 | |||

IL17RA
Genomic Map