siRNA Details
| Gene Symbol: INVS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001318381.1 | NP_001305310.1 | 8 | 1182 | ||
| NM_001318382.1 | NP_001305311.1 | 7 | 1094 | |||
| NM_014425.4 | NP_055240.2 | 7 | 1094 | |||
| NR_134606.1 | 7 | 1094 | ||||
| GenBank mRNA | AF039217.2 | - | 7 | 1045 | ||
| AF084367.1 | - | 6 | 864 | |||
| AK304851.1 | - | 8 | 1104 | |||
| AK312795.1 | - | 7 | 1036 | |||
| BC041665.1 | - | 7 | 1050 | |||
| BC056897.1 | - | 9 | 1217 | |||
| BC111761.1 | - | 6 | 877 | 40012263 | ||
| BC112351.1 | - | 6 | 877 | 40012265 | ||

INVS
Genomic Map