siRNA Details
| Gene Symbol: LPIN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014646.2 | NP_055461.1 | 10 | 1755 | ||
| XM_005258177.4 | XP_005258234.1 | 10 | 1825 | |||
| XM_005258178.3 | XP_005258235.1 | 10 | 1754 | |||
| XM_005258179.4 | XP_005258236.1 | 10 | 1688 | |||
| XM_017026098.1 | XP_016881587.1 | 11 | 1844 | |||
| XM_017026099.1 | XP_016881588.1 | 10 | 1615 | |||
| GenBank mRNA | BC152448.1 | - | 10 | 1755 | 8862579 | |
| D87436.1 | - | 10 | 1755 | |||

LPIN2
Genomic Map