siRNA Details
| Gene Symbol: MGME1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001310338.1 | NP_001297267.1 | 2 | 327 | ||
| NM_001310339.1 | NP_001297268.1 | 2 | 327 | |||
| NM_052865.3 | NP_443097.1 | 2 | 327 | |||
| XM_005260867.2 | XP_005260924.1 | 2 | 549 | |||
| XM_005260870.3 | XP_005260927.1 | 2 | 275 | |||
| XM_006723663.3 | XP_006723726.1 | 2 | 275 | |||
| XM_011529394.1 | XP_011527696.1 | 2 | 164 | |||
| XM_011529395.1 | XP_011527697.1 | 2 | 327 | |||
| XM_017028127.1 | XP_016883616.1 | 2 | 549 | |||
| XM_017028128.1 | XP_016883617.1 | 2 | 423 | |||
| GenBank mRNA | AF306684.1 | - | 2 | 135 | ||
| AF306685.1 | - | 2 | 135 | |||
| AF306686.1 | - | 2 | 173 | |||
| AK027503.1 | - | 2 | 131 | |||
| AK315532.1 | - | 2 | 163 | |||
| BC016869.1 | - | 2 | 99 | 3850853 | ||
| BX382492.2 | - | 1 | 92 | |||
| CV807140.1 | - | 2 | 140 | |||
| DB014120.1 | - | 2 | 327 | |||

MGME1
Genomic Map