siRNA Details
| Gene Symbol: TTC19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271420.1 | NP_001258349.1 | 7 | 1228 | ||
| NM_017775.3 | NP_060245.3 | 7 | 1091 | |||
| XM_017024801.1 | XP_016880290.1 | 7 | 1004 | |||
| XM_017024802.1 | XP_016880291.1 | 7 | 1004 | |||
| GenBank mRNA | AK000350.1 | - | 5 | 307 | ||
| AK055780.1 | - | 7 | 784 | |||
| AK056878.1 | - | 7 | 1003 | |||
| AK297783.1 | - | 7 | 647 | |||
| BC011698.2 | - | 7 | 630 | |||
| BC073796.1 | - | 7 | 634 | |||
| BC105128.1 | - | 7 | 1091 | 8144131 | ||
| BC112107.1 | - | 7 | 1091 | 8327575 | ||

TTC19
Genomic Map