siRNA Details
| Gene Symbol: OBSL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001173408.1 | NP_001166879.1 | 9 | 3287 | ||
| GenBank mRNA | AF035292.1 | - | 6 | 1413 | ||
| AK025946.1 | - | 8 | 1740 | |||
| BC007201.1 | - | 9 | 3032 | |||
| BC061909.1 | - | 4 | 643 | |||

OBSL1
Genomic Map