siRNA Details
| Gene Symbol: RSPH3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031924.4 | NP_114130.3 | 2 | 782 | ||
| XM_005267153.4 | XP_005267210.1 | 2 | 1048 | |||
| XM_017011347.1 | XP_016866836.1 | 2 | 245 | |||
| GenBank mRNA | AF353618.1 | - | 2 | 381 | ||
| AK057655.1 | - | 2 | 397 | |||
| AK057931.1 | - | 2 | 803 | |||
| BC050604.1 | - | 2 | 782 | 6161669 | ||

RSPH3
Genomic Map