siRNA Details
| Gene Symbol: SERTM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_203451.2 | NP_982276.2 | 2 | 1244 | ||
| GenBank mRNA | BC036540.2 | - | 2 | 1244 | 5260372 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

SERTM1| Gene Symbol: SERTM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_203451.2 | NP_982276.2 | 2 | 1244 | ||
| GenBank mRNA | BC036540.2 | - | 2 | 1244 | 5260372 | |