siRNA Details
| Gene Symbol: RSPO4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001029871.3 | NP_001025042.2 | 1 | 163 | ||
| NM_001040007.2 | NP_001035096.1 | 1 | 163 | |||
| XM_017027839.1 | XP_016883328.1 | 1 | 91 | |||
| GenBank mRNA | AK122609.1 | - | 1 | 160 | ||
| DQ355152.1 | - | 1 | 163 | |||

RSPO4
Genomic Map