siRNA Details
| Gene Symbol: FAM20A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001243746.1 | NP_001230675.1 | 12 | 4133 | ||
| NM_017565.3 | NP_060035.2 | 11 | 3907 | |||
| NR_027751.1 | 11 | 3334 | ||||
| XM_006721959.2 | XP_006722022.1 | 11 | 3279 | |||
| GenBank mRNA | AB545606.1 | - | 11 | 3907 | ||
| BC036222.1 | - | 11 | 3300 | |||

FAM20A
Genomic Map