siRNA Details
| Gene Symbol: CYBA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000101.3 | NP_000092.2 | 2 | 190 | ||
| XM_011522905.2 | XP_011521207.1 | 2 | 185 | |||
| GenBank mRNA | AK097127.1 | - | 2 | 93 | ||
| AK300049.1 | - | 2 | 94 | |||
| BC006465.1 | - | 2 | 146 | 3502788 | ||
| BT006861.1 | - | 2 | 119 | |||
| BX109994.1 | - | 2 | 155 | |||
| M21186.1 | - | 2 | 147 | |||

CYBA
Genomic Map