siRNA Details
| Gene Symbol: CYBA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000101.3 | NP_000092.2 | 4 | 334 | ||
| XM_011522905.2 | XP_011521207.1 | 4 | 329 | |||
| GenBank mRNA | AK097127.1 | - | 4 | 237 | ||
| AK300049.1 | - | 4 | 238 | |||
| AK309030.1 | - | 4 | 238 | |||
| BC006465.1 | - | 4 | 290 | 3502788 | ||
| BT006861.1 | - | 4 | 263 | |||
| BX109994.1 | - | 4 | 299 | |||
| M21186.1 | - | 4 | 291 | |||

CYBA
Genomic Map