siRNA Details
| Gene Symbol: IARS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018060.3 | NP_060530.3 | 6 | 901 | ||
| GenBank mRNA | AK001188.1 | - | 4 | 376 | ||
| AK291432.1 | - | 6 | 710 | |||
| AY267462.1 | - | 6 | 912 | |||
| BC010218.1 | - | 2 | 81 | |||
| BC040376.1 | - | 6 | 634 | |||
| BC047880.1 | - | 5 | 428 | |||
| BC137438.1 | - | 6 | 871 | 9021436 | ||
| D28500.1 | - | 6 | 742 | |||

IARS2
Genomic Map