siRNA Details
| Gene Symbol: PLEKHD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001161498.1 | NP_001154970.1 | 5, 6 | 709 | ||
| XM_011536762.2 | XP_011535064.1 | 2, 3 | 155 | |||
| XM_017021290.1 | XP_016876779.1 | 5, 6 | 1071 | |||
| GenBank mRNA | BC146872.1 | - | 5, 6 | 614 | 9056800 | |

PLEKHD1
Genomic Map