siRNA Details
| Gene Symbol: RASA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_002890.2 | NP_002881.1 | 5 | 1157 | ||
| NM_022650.2 | NP_072179.1 | 5 | 551 | |||
| XM_011543525.1 | XP_011541827.1 | 5 | 1609 | |||
| XM_011543527.2 | XP_011541829.1 | 5 | 1609 | |||
| GenBank mRNA | AB209106.1 | - | 5 | 1157 | ||
| AK300263.1 | - | 6 | 658 | |||
| AK300269.1 | - | 5 | 484 | |||
| AK300406.1 | - | 6 | 577 | |||
| AK312739.1 | - | 5 | 536 | |||
| BC020761.1 | - | 5 | 538 | |||
| BC033015.2 | - | 5 | 1145 | 4829173 | ||
| BC054891.1 | - | 5 | 517 | |||
| BC107870.1 | - | 5 | 541 | |||
| CR749722.1 | - | 5 | 1522 | |||
| DA863496.1 | - | 5 | 552 | |||
| M23379.1 | - | 5 | 1076 | |||
| M23612.1 | - | 5 | 526 | |||

RASA1
Genomic Map