siRNA Details
| Gene Symbol: WDR34 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_052844.3 | NP_443076.2 | 2, 3 | 564 | ||
| XM_011519179.2 | XP_011517481.1 | 2, 3 | 498 | |||
| GenBank mRNA | AL555205.3 | - | 2, 3 | 371 | ||
| BC001614.1 | - | 2, 3 | 399 | |||
| BC011874.2 | - | 2, 3 | 501 | 3161070 | ||
| BG177634.1 | - | 2, 3 | 408 | |||
| BI910783.1 | - | 3, 4 | 659 | |||

WDR34
Genomic Map