siRNA Details
| Gene Symbol: SH2D2A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001161441.1 | NP_001154913.1 | 6 | 825 | ||
| NM_001161442.1 | NP_001154914.1 | 6 | 764 | |||
| NM_001161443.1 | NP_001154915.1 | 5 | 674 | |||
| NM_001161444.1 | NP_001154916.1 | 6 | 795 | |||
| NM_003975.3 | NP_003966.2 | 6 | 795 | |||
| XM_006711615.2 | XP_006711678.1 | 6 | 813 | |||
| XM_017002762.1 | XP_016858251.1 | 6 | 842 | |||
| XM_017002763.1 | XP_016858252.1 | 6 | 833 | |||
| XM_017002764.1 | XP_016858253.1 | 6 | 780 | |||
| XM_017002765.1 | XP_016858254.1 | 6 | 793 | |||
| XM_017002766.1 | XP_016858255.1 | 6 | 783 | |||
| XM_017002767.1 | XP_016858256.1 | 6 | 848 | |||
| GenBank mRNA | AF051325.1 | - | 6 | 717 | ||
| AF097744.1 | - | 6 | 694 | |||
| AJ000553.1 | - | 6 | 741 | |||
| AK222737.1 | - | 6 | 718 | |||
| AK313706.1 | - | 6 | 789 | |||
| AY763098.1 | - | 5 | 674 | |||
| AY763099.1 | - | 6 | 771 | |||
| AY763100.1 | - | 6 | 701 | |||
| BC012107.1 | - | 6 | 757 | 4636736 | ||

SH2D2A
Genomic Map