siRNA Details
| Gene Symbol: SLC27A4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005094.3 | NP_005085.2 | 13 | 2083 | ||
| XM_017014222.1 | XP_016869711.1 | 14 | 1956 | |||
| GenBank mRNA | AF055899.1 | - | 12 | 1795 | ||
| AK027504.1 | - | 12 | 1663 | |||
| AK074831.1 | - | 10 | 1131 | |||
| AK290222.1 | - | 13 | 2083 | |||
| AK314681.1 | - | 12 | 2103 | |||
| BC004268.1 | - | 6 | 643 | |||
| BC009959.2 | - | 6 | 696 | |||
| BC065003.1 | - | 13 | 2082 | 6023438 | ||

SLC27A4
Genomic Map