siRNA Details
| Gene Symbol: SLC27A4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005094.3 | NP_005085.2 | 12, 13 | 2052 | ||
| XM_017014222.1 | XP_016869711.1 | 13, 14 | 1925 | |||
| GenBank mRNA | AF055899.1 | - | 11, 12 | 1764 | ||
| AK000722.1 | - | 3, 4 | 414 | |||
| AK027504.1 | - | 11, 12 | 1632 | |||
| AK074831.1 | - | 9, 10 | 1100 | |||
| AK290222.1 | - | 12, 13 | 2052 | |||
| AK314681.1 | - | 11, 12 | 2072 | |||
| BC004268.1 | - | 5, 6 | 612 | |||
| BC009959.2 | - | 5, 6 | 665 | |||
| BC065003.1 | - | 12, 13 | 2051 | 6023438 | ||

SLC27A4
Genomic Map