siRNA Details
| Gene Symbol: CORO1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001193333.2 | NP_001180262.1 | 6, 7 | 1069 | ||
| NM_007074.3 | NP_009005.1 | 5, 6 | 951 | |||
| XM_011545714.2 | XP_011544016.1 | 5, 6 | 764 | |||
| XM_017022885.1 | XP_016878374.1 | 5, 6 | 939 | |||
| XM_017022886.1 | XP_016878375.1 | 5, 6 | 761 | |||
| GenBank mRNA | AB209221.1 | - | 2, 3 | 2896 | ||
| AF495470.1 | - | 5, 6 | 756 | |||
| AK096332.1 | - | 4, 5 | 1946 | |||
| AK097964.1 | - | 1, 2 | 540 | |||
| AK123401.1 | - | 2, 3 | 1195 | |||
| AK314714.1 | - | 6, 7 | 870 | |||
| BC110374.1 | - | 5, 6 | 768 | 5733853 | ||
| BC126385.1 | - | 5, 6 | 655 | 8992101 | ||
| BC126387.1 | - | 5, 6 | 655 | 8992103 | ||
| D44497.1 | - | 5, 6 | 734 | |||
| U34690.1 | - | 5, 6 | 726 | |||
| X89109.1 | - | 5, 6 | 727 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BC064330.1 | - | 2411 | Chr. - - [-] on Build | ||

CORO1A
Genomic Map