siRNA Details
| Gene Symbol: PAX9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006194.3 | NP_006185.1 | 4, 5 | 1495 | ||
| GenBank mRNA | AB248958.1 | - | 3, 4 | 938 | ||
| AK313627.1 | - | 3, 4 | 950 | |||
| BC001159.1 | - | 3, 4 | 958 | 3534582 | ||
| U59628.1 | - | 2, 3 | 629 | |||
| X92850.1 | - | 2, 3 | 745 | |||

PAX9
Genomic Map