siRNA Details
| Gene Symbol: PAX9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006194.3 | NP_006185.1 | 5 | 1578 | ||
| GenBank mRNA | AB248958.1 | - | 4 | 1021 | ||
| AK310339.1 | - | 5 | 463 | |||
| AK313627.1 | - | 4 | 1033 | |||
| BC001159.1 | - | 4 | 1041 | 3534582 | ||
| U59628.1 | - | 3 | 712 | |||
| X92850.1 | - | 3 | 828 | |||

PAX9
Genomic Map