siRNA Details
| Gene Symbol: PAX9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006194.3 | NP_006185.1 | 3 | 771 | ||
| GenBank mRNA | AB248958.1 | - | 2 | 214 | ||
| AK313627.1 | - | 2 | 226 | |||
| BC001159.1 | - | 2 | 234 | 3534582 | ||
| X92850.1 | - | 1 | 21 | |||

PAX9
Genomic Map