siRNA Details
Gene Symbol: KRT6A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005554.3 | NP_005545.1 | 9 | 2417 | ||
GenBank mRNA | BC008807.2 | - | 9 | 2256 | 3947610 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: KRT6A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005554.3 | NP_005545.1 | 9 | 2417 | ||
GenBank mRNA | BC008807.2 | - | 9 | 2256 | 3947610 |