siRNA Details
| Gene Symbol: KNOP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012991.2 | NP_001013009.2 | 4 | 1351 | ||
| XM_005255317.4 | XP_005255374.1 | 6 | 1546 | |||
| XM_006721046.3 | XP_006721109.1 | 5 | 1709 | |||
| XM_011545855.1 | XP_011544157.1 | 5 | 1595 | |||
| XM_017023231.1 | XP_016878720.1 | 5 | 1487 | |||
| XM_017023232.1 | XP_016878721.1 | 4 | 1632 | |||
| GenBank mRNA | BC117562.1 | - | 4 | 1351 | 7971177 | |

KNOP1
Genomic Map