siRNA Details
| Gene Symbol: G6PC3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001319945.1 | NP_001306874.1 | 5 | 1402 | ||
| NM_138387.3 | NP_612396.1 | 6 | 1521 | |||
| NR_028581.1 | 7 | 1715 | ||||
| NR_028582.1 | 6 | 1580 | ||||
| XM_011525473.2 | XP_011523775.1 | 7 | 1275 | |||
| XM_011525474.2 | XP_011523776.1 | 7 | 1744 | |||
| XM_017025335.1 | XP_016880824.1 | 6 | 1266 | |||
| GenBank mRNA | BC002494.2 | - | 6 | 1319 | 3050476 | |
| BC021574.1 | - | 6 | 1501 | 4842511 | ||

G6PC3
Genomic Map