siRNA Details
| Gene Symbol: DNAI2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001172810.1 | NP_001166281.1 | 10 | 1409 | ||
| NM_023036.4 | NP_075462.3 | 10 | 1412 | |||
| XM_011525125.2 | XP_011523427.1 | 10 | 1429 | |||
| GenBank mRNA | AF250288.1 | - | 10 | 1369 | ||
| AJ295276.1 | - | 9 | 1608 | |||
| AL137526.1 | - | 2 | 355 | |||
| BC033013.1 | - | 9 | 1286 | 4826432 | ||
| BC039582.1 | - | 10 | 1399 | 5752153 | ||

DNAI2
Genomic Map