siRNA Details
| Gene Symbol: DNAI2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001172810.1 | NP_001166281.1 | 14 | 2049 | ||
| NM_023036.4 | NP_075462.3 | 14 | 2088 | |||
| XM_011525125.2 | XP_011523427.1 | 15 | 2212 | |||
| GenBank mRNA | AJ295276.1 | - | 13 | 2284 | ||
| AL137526.1 | - | 6 | 1031 | |||
| BC033013.1 | - | 13 | 1807 | 4826432 | ||
| BC039582.1 | - | 14 | 2039 | 5752153 | ||
| DB514781.1 | - | 2 | 228 | |||

DNAI2
Genomic Map