siRNA Details
| Gene Symbol: ITK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005546.3 | NP_005537.3 | 4, 5 | 536 | ||
| XM_017009443.1 | XP_016864932.1 | 2, 3 | 149 | |||
| GenBank mRNA | AB209622.1 | - | 4, 5 | 589 | ||
| AK312846.1 | - | 4, 5 | 505 | |||
| BC109077.2 | - | 4, 5 | 527 | 40034274 | ||
| BC109078.1 | - | 4, 5 | 527 | 40034276 | ||
| D13720.1 | - | 4, 5 | 536 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | L10717.1 | - | 2477 | Chr. - - [-] on Build | ||

ITK
Genomic Map